siRNA Details
| Gene Symbol: Ddx49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115023.1 | NP_001108495.1 | 12 | 1235 | ||
| GenBank mRNA | BC158855.1 | - | 12 | 1235 | 7930984 | |
| FQ216881.1 | - | 10 | 2534 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddx49| Gene Symbol: Ddx49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115023.1 | NP_001108495.1 | 12 | 1235 | ||
| GenBank mRNA | BC158855.1 | - | 12 | 1235 | 7930984 | |
| FQ216881.1 | - | 10 | 2534 | |||