siRNA Details
| Gene Symbol: Pcp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107116.1 | NP_001100586.1 | 3 | 227 | ||
| XM_006248762.2 | XP_006248824.1 | 3 | 297 | |||
| XM_008768953.1 | XP_008767175.1 | 3 | 282 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcp2| Gene Symbol: Pcp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107116.1 | NP_001100586.1 | 3 | 227 | ||
| XM_006248762.2 | XP_006248824.1 | 3 | 297 | |||
| XM_008768953.1 | XP_008767175.1 | 3 | 282 | |||