siRNA Details
| Gene Symbol: Xpo7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108386.2 | NP_001101856.2 | 2 | 157 | ||
| XM_006252282.2 | XP_006252344.1 | 2 | 1643 | |||
| XM_008770832.1 | XP_008769054.1 | 2 | 1686 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xpo7| Gene Symbol: Xpo7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108386.2 | NP_001101856.2 | 2 | 157 | ||
| XM_006252282.2 | XP_006252344.1 | 2 | 1643 | |||
| XM_008770832.1 | XP_008769054.1 | 2 | 1686 | |||