siRNA Details
| Gene Symbol: LOC302022 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127530.1 | NP_001121002.1 | 285 | |||
| GenBank mRNA | BC162000.1 | - | 285 | 9036094 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC302022| Gene Symbol: LOC302022 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127530.1 | NP_001121002.1 | 285 | |||
| GenBank mRNA | BC162000.1 | - | 285 | 9036094 | ||