This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CITED4 SIRNA Assays ›
Gene Symbol:
Cited4
Gene Name:
Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 4
Gene Aliases:
MRG-2, Mrg2
Chromosome Location:
Chr.5: 139597731 - 139598622 on Build Rnor_6.0
UniGene ID:
Rn.40380
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s236370

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cited4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053699.1 NP_446151.1 1 745
GenBank mRNA AF361477.1 - 1 745

Back To Top

More Information


Panther Classification:

Molecular Function -

transcription cofactor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
response to estrogen
positive regulation of transcription, DNA-templated
regulation of nucleic acid-templated transcription
transcription coactivator activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-ztldj:80/100.66.73.158:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline