siRNA Details
| Gene Symbol: Brdt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012031.2 | NP_001012031.1 | 7 | 1210 | ||
| GenBank mRNA | BC078999.1 | - | 7 | 1188 | ||
| BC158630.1 | - | 7 | 1185 | 7456383 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Brdt| Gene Symbol: Brdt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012031.2 | NP_001012031.1 | 7 | 1210 | ||
| GenBank mRNA | BC078999.1 | - | 7 | 1188 | ||
| BC158630.1 | - | 7 | 1185 | 7456383 | ||