This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other KLHL23 SIRNA Assays ›
Gene Symbol:
Klhl23
Gene Name:
kelch-like family member 23
Gene Aliases:
RGD1307166
Chromosome Location:
Chr.3: 56057043 - 56070614 on Build Rnor_6.0
UniGene ID:
Rn.125724
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s236679

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 296.00
296.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Klhl23
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134504.1 NP_001127976.1 5 2387
XM_006234313.2 XP_006234375.1 4 3259
XM_006234314.2 XP_006234376.1 4 3755
XM_006234316.2 XP_006234378.1 4 3500
GenBank mRNA FQ217393.1 - 1 737

Back To Top

More Information


Panther Classification:

Molecular Function -

scaffold/adaptor protein

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-smrv7:80/100.66.72.156:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0