This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FAM227A SIRNA Assays ›
Gene Symbol:
Fam227a
Gene Name:
family with sequence similarity 227, member A
Gene Aliases:
RGD1305939
Chromosome Location:
Chr.7: 120846166 - 120891765 on Build Rnor_6.0
UniGene ID:
Rn.72811
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s236798

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 404.00
Your Price
Online offer: 391.65
391.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fam227a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008765683.1 XP_008763905.1 10 1392
GenBank mRNA BC097470.1 - 11 1193 7457750
CB546465.1 - 3 259

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline