This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other HMGB2 SIRNA Assays ›
Gene Symbol:
Hmgb2 Hmgb2l1
Gene Name:
high mobility group box 2
high mobility group box 2-like 1
Gene Aliases:
Hmg2
RGD1564519
Chromosome Location:
Chr.16: 36077617 - 36080191 on Build Rnor_6.0
Chromosome Location:
Chr.11: 49708042 - 49709128 on Build Rnor_6.0
UniGene ID:
Rn.218982
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s236958

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hmgb2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017187.2 NP_058883.1 5 1071
GenBank mRNA BC078866.1 - 4 930 7114119
BC089854.1 - 5 1071 7380208
BC107455.1 - 5 1100 7377505
FQ225084.1 - 5 1035
FQ229618.1 - 5 1005
FQ230440.1 - 5 1035
FQ233747.1 - 5 1008
Gene Symbol: Hmgb2l1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_573272.5 XP_573272.3 1 1009

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008252, AK003773, AK076075, BC083108, AK135296, and others.

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of endothelial cell proliferation
base-excision repair, DNA ligation
chromatin remodeling
regulation of transcription from RNA polymerase II promoter
spermatogenesis
spermatid nucleus differentiation
nervous system development
male gonad development
organ regeneration
positive regulation of nuclease activity
positive regulation of DNA binding
positive regulation of erythrocyte differentiation
positive regulation of megakaryocyte differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
response to steroid hormone
positive chemotaxis
positive regulation of DNA metabolic process
cell chemotaxis
cellular response to lipopolysaccharide
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
biological_process
DNA binding
damaged DNA binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
lipid binding
DNA binding, bending
protein domain specific binding
nucleosomal histone binding
chemoattractant activity
transcription regulatory region DNA binding
poly(A) RNA binding
RAGE receptor binding
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-8pkvz:80/100.66.73.30:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0