siRNA Details
| Gene Symbol: MKRN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014160.4 | NP_054879.3 | 2 | 243 | ||
| GenBank mRNA | AF302084.1 | - | 2 | 157 | ||
| AK091318.1 | - | 2 | 188 | |||
| BC015715.1 | - | 2 | 185 | 3916199 | ||
| DR157266.1 | - | 2 | 243 | |||

MKRN2
Genomic Map