This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TMEM184B SIRNA Assays ›
Gene Symbol:
TMEM184B
Gene Name:
transmembrane protein 184B
Gene Aliases:
C22orf5, FM08, HS5O6A, HSPC256
Chromosome Location:
Chr.22: 38216045 - 38273034 on Build GRCh38
UniGene ID:
Hs.182626
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s24571

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TMEM184B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195071.1 NP_001182000.1 5 720
NM_001195072.1 NP_001182001.1 6 806
NM_012264.4 NP_036396.2 5 705
XM_011530112.2 XP_011528414.2 5 754
XM_011530113.1 XP_011528415.1 5 684
XM_011530114.1 XP_011528416.1 5 716
XM_011530115.2 XP_011528417.1 4 475
XM_011530116.2 XP_011528418.2 5 746
XM_011530117.1 XP_011528419.1 6 782
XM_011530118.1 XP_011528420.1 6 756
XM_011530119.1 XP_011528421.1 4 510
XM_017028754.1 XP_016884243.1 5 755
XM_017028755.1 XP_016884244.1 5 748
XM_017028756.1 XP_016884245.1 5 684
XM_017028757.1 XP_016884246.1 4 472
XM_017028758.1 XP_016884247.1 5 684
XM_017028759.1 XP_016884248.1 4 510
GenBank mRNA AB097053.1 - 5 676
AK075417.1 - 3 193
AK292652.1 - 5 705
AK309071.1 - 5 741
AL096879.1 - 4 398
BC012983.2 - 1 29
BC015489.1 - 5 661 3896682
BI197128.1 - 4 399
BX648345.1 - 5 756
CR456413.1 - 4 539
CR749859.1 - 6 712

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001173370, XM_006242013, XM_006242014, BC099785.

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-2x92s:80/100.66.79.168:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0