This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF473 SIRNA Assays ›
Gene Symbol:
ZNF473
Gene Name:
zinc finger protein 473
Gene Aliases:
ZFP100, ZN473
Chromosome Location:
Chr.19: 50025806 - 50048776 on Build GRCh38
UniGene ID:
Hs.440553
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s24680

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF473
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017026574.1 XP_016882063.1 3 2171
GenBank mRNA AB032967.1 - 5 928
AF454744.1 - 4 883
AK054671.1 - 1 70
AK292452.1 - 5 821
AK302293.1 - 4 778
BC018612.1 - 5 851 4634056
BX648093.1 - 5 972

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
termination of RNA polymerase II transcription
mRNA 3'-end processing by stem-loop binding and cleavage
histone mRNA metabolic process
DNA binding
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-gdckb:80/100.66.73.39:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0