This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FBXL5 SIRNA Assays ›
Gene Symbol:
FBXL5
Gene Name:
F-box and leucine rich repeat protein 5
Gene Aliases:
FBL4, FBL5, FLR1
Chromosome Location:
Chr.4: 15604384 - 15681545 on Build GRCh38
UniGene ID:
Hs.643433
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s25273

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FBXL5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193534.1 NP_001180463.1 9 1658
NM_001193535.1 NP_001180464.1 9 1610
NM_012161.3 NP_036293.1 9 1661
NR_036464.1 10 1765
XM_006713959.3 XP_006714022.1 10 1757
XM_011513831.2 XP_011512133.1 9 1443
XM_011513832.2 XP_011512134.1 9 1590
XM_011513833.2 XP_011512135.1 8 1401
XM_017008019.1 XP_016863508.1 11 2528
GenBank mRNA AF052097.1 - 1 220
AF142481.1 - 9 1583
AF157323.1 - 9 1522
AF174591.1 - 6 1068
AF176700.1 - 9 1545
AF199420.1 - 9 1547
AK000153.1 - 8 1371
AK295510.1 - 9 1580
AK310860.1 - 9 1590
AL832808.1 - 7 1297
BC012320.1 - 10 1694
BC030656.1 - 9 1632 4837080
BX537957.1 - 9 1556

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
iron ion homeostasis
positive regulation of cellular protein catabolic process
ubiquitin-protein transferase activity
iron ion binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-jq955:80/100.66.72.86:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0