siRNA Details
| Gene Symbol: FBXW8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012174.1 | NP_036306.1 | 7 | 1108 | ||
| NM_153348.2 | NP_699179.2 | 7 | 1306 | |||
| XM_005253867.2 | XP_005253924.1 | 6 | 1164 | |||
| XM_005253868.3 | XP_005253925.1 | 6 | 997 | |||
| XM_017019175.1 | XP_016874664.1 | 7 | 1083 | |||
| XM_017019176.1 | XP_016874665.1 | 8 | 1315 | |||
| XM_017019177.1 | XP_016874666.1 | 8 | 1315 | |||
| XM_017019178.1 | XP_016874667.1 | 3 | 2057 | |||
| GenBank mRNA | AF176707.1 | - | 7 | 1083 | ||
| AK289621.1 | - | 7 | 1099 | |||
| BC037296.2 | - | 7 | 1306 | 4821240 | ||

FBXW8
Genomic Map