This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other IL1RAPL2 SIRNA Assays ›
Gene Symbol:
IL1RAPL2
Gene Name:
interleukin 1 receptor accessory protein like 2
Gene Aliases:
IL-1R9, IL1R9, IL1RAPL-2, TIGIRR-1
Chromosome Location:
Chr.X: 104566315 - 105769729 on Build GRCh38
UniGene ID:
Hs.675519
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s25366

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: IL1RAPL2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017416.1 NP_059112.1 5 1422
XM_011530905.2 XP_011529207.1 3 430
XM_017029403.1 XP_016884892.1 5 840
GenBank mRNA AF181285.1 - 3 584
AF212016.1 - 4 666
AF284436.1 - 5 817
AJ272208.1 - 5 1422
AJ290436.1 - 4 685
AK290196.1 - 5 1538
BC104784.1 - 5 714 8143787
BC104786.1 - 5 714 8143789

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_030688, BC120555, BC119274, AK039155, AF284437, and others.

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

central nervous system development
cytokine-mediated signaling pathway
interleukin-1 receptor activity
interleukin-1, Type II, blocking receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gm74s:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline