This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other OR8G1 SIRNA Assays ›
Gene Symbol:
OR8G1
Gene Name:
olfactory receptor family 8 subfamily G member 1 (gene/pseudogene)
Gene Aliases:
HSTPCR25, OR8G1P, TPCR25
Chromosome Location:
Chr.11: 124249676 - 124250611 on Build GRCh38
UniGene ID:
Hs.448172
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s25450

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 388.00
Your Price
Online offer: 374.65
374.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: OR8G1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002905.1 NP_001002905.1 1 578
NR_045681.1 1 578
GenBank mRNA X89672.1 - 1 204

Back To Top

More Information


Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
sensory perception of smell
detection of chemical stimulus involved in sensory perception of smell
G-protein coupled receptor activity
olfactory receptor activity
odorant binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-h5txz:80/100.66.79.227:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline