This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TSPAN16 SIRNA Assays ›
Gene Symbol:
TSPAN16
Gene Name:
tetraspanin 16
Gene Aliases:
TM-8, TM4-B, TM4SF16
Chromosome Location:
Chr.19: 11296139 - 11326996 on Build GRCh38
UniGene ID:
Hs.579784
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s25510

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TSPAN16
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282509.1 NP_001269438.1 4 595
NM_001282510.1 NP_001269439.1 3 520
NM_012466.3 NP_036598.1 4 595
NR_104199.1 4 595
XM_011527901.1 XP_011526203.1 4 590
XM_011527903.2 XP_011526205.1 4 526
GenBank mRNA BC029908.1 - 4 565 5168677
BM560276.1 - 4 582

Back To Top

More Information


Gene Ontology Categories:

Function(s)

cell surface receptor signaling pathway

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0