siRNA Details
| Gene Symbol: RNF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014372.4 | NP_055187.1 | 2 | 754 | ||
| GenBank mRNA | AB024703.1 | - | 2 | 397 | ||
| AF151881.1 | - | 2 | 397 | |||
| AK293047.1 | - | 4 | 583 | |||
| AK313140.1 | - | 2 | 516 | |||
| BC020964.2 | - | 2 | 349 | 3917224 | ||
| BC047654.1 | - | 2 | 722 | 5261966 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_013876, AF220206, AB024427, BC010299, AK014254, and others.

RNF11
Genomic Map