This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PKD2L2 SIRNA Assays ›
Gene Symbol:
PKD2L2
Gene Name:
polycystin 2 like 2, transient receptor potential cation channel
Gene Aliases:
TRPP5
Chromosome Location:
Chr.5: 137889420 - 137942747 on Build GRCh38
UniGene ID:
Hs.716884
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s25737

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 388.00
Your Price
Online offer: 287.12
287.12
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PKD2L2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258448.1 NP_001245377.1 12 1650
NM_001258449.1 NP_001245378.1 10 1413
NM_001300921.1 NP_001287850.1 12 1716
NM_014386.3 NP_055201.2 12 1716
XM_011543318.2 XP_011541620.1 11 1630
XM_011543321.1 XP_011541623.1 12 1650
XM_017009343.1 XP_016864832.1 12 1732
XM_017009344.1 XP_016864833.1 11 1840
XM_017009345.1 XP_016864834.1 11 1510
GenBank mRNA AF118125.1 - 12 1683
AF182034.1 - 12 1686
AK301924.1 - 12 1619
BC044581.1 - 10 1412 4830805

Back To Top

More Information


Panther Classification:

Molecular Function -

ion channel

Gene Ontology Categories:

Function(s) Process(es)

biological_process
detection of mechanical stimulus
calcium ion transmembrane transport
calcium channel activity
calcium ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-x4bxv:80/100.66.79.138:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline