siRNA Details
| Gene Symbol: TMEM97 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014573.2 | NP_055388.2 | 3 | 613 | ||
| XM_005257965.3 | XP_005258022.1 | 3 | 662 | |||
| GenBank mRNA | AK293331.1 | - | 3 | 468 | ||
| AK299511.1 | - | 3 | 506 | |||
| AK307678.1 | - | 3 | 505 | |||
| BC017362.2 | - | 3 | 484 | |||
| BC045655.1 | - | 4 | 622 | |||
| BC091504.1 | - | 3 | 484 | 30379019 | ||
| L19183.1 | - | 3 | 507 | |||

TMEM97
Genomic Map