siRNA Details
| Gene Symbol: ZNF354C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014594.2 | NP_055409.1 | 5 | 1837 | ||
| XM_017009409.1 | XP_016864898.1 | 5 | 1940 | |||
| GenBank mRNA | AF395540.1 | - | 3 | 1504 | ||
| AF525463.1 | - | 5 | 1646 | |||
| AL137483.1 | - | 1 | 827 | |||
| BC063312.1 | - | 5 | 1835 | 30341631 | ||

ZNF354C
Genomic Map