This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other BOLA1 SIRNA Assays ›
Gene Symbol:
BOLA1
Gene Name:
bolA family member 1
Gene Aliases:
CGI-143
Chromosome Location:
Chr.1: 149899582 - 149900795 on Build GRCh38
UniGene ID:
Hs.13880
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s27274

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 388.00
Your Price
Online offer: 287.12
287.12
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: BOLA1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321025.1 NP_001307954.1 2 962
NM_001321026.1 NP_001307955.1 2 1020
NM_016074.4 NP_057158.1 2 795
XM_006711348.3 XP_006711411.1 2 828
GenBank mRNA AF151901.1 - 2 731
BC002771.2 - 2 755
BC063405.1 - 2 755 5190565
BQ277416.1 - 1 161

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline