This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF580 SIRNA Assays ›
Gene Symbol:
ZNF580
Gene Name:
zinc finger protein 580
Gene Aliases:
-
Chromosome Location:
Chr.19: 55641026 - 55643470 on Build GRCh38
UniGene ID:
Hs.631551
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s27575

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 388.00
Your Price
Online offer: 287.12
287.12
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF580
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163423.1 NP_001156895.1 2 953
NM_016202.2 NP_057286.1 1 1346
NM_207115.1 NP_996998.1 2 1059
GenBank mRNA AF184939.1 - 1 1633
AL359054.1 - 2 1149
BC017698.1 - 1 1143 4431902

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of endothelial cell proliferation
positive regulation of leukocyte chemotaxis
transcription, DNA-templated
regulation of transcription, DNA-templated
chemotaxis
inflammatory response
positive regulation of endothelial cell migration
positive regulation of gene expression
positive regulation of interleukin-8 production
cellular response to hydrogen peroxide
DNA binding
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-wtj7v:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline