siRNA Details
| Gene Symbol: HSD17B14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016246.2 | NP_057330.2 | 9 | 1169 | ||
| XM_005258969.3 | XP_005259026.1 | 8 | 1081 | |||
| XM_005258970.3 | XP_005259027.1 | 7 | 991 | |||
| GenBank mRNA | AY358430.1 | - | 9 | 1008 | ||
| BC006283.2 | - | 9 | 962 | 4026397 | ||
| BC006294.1 | - | 9 | 970 | 4025809 | ||

HSD17B14
Genomic Map