This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PHF20 SIRNA Assays ›
Gene Symbol:
PHF20
Gene Name:
PHD finger protein 20
Gene Aliases:
C20orf104, GLEA2, HCA58, NZF, TDRD20A, TZP
Chromosome Location:
Chr.20: 35771999 - 35950377 on Build GRCh38
UniGene ID:
Hs.517044
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s27721

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PHF20
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016436.4 NP_057520.2 6, 7 922
XM_005260421.2 XP_005260478.1 3, 4 532
XM_011528844.1 XP_011527146.1 3, 4 594
XM_017027864.1 XP_016883353.1 7, 8 1004
XM_017027865.1 XP_016883354.1 7, 8 1205
XM_017027866.1 XP_016883355.1 6, 7 1125
XM_017027867.1 XP_016883356.1 6, 7 960
XM_017027868.1 XP_016883357.1 4, 5 746
XM_017027869.1 XP_016883358.1 6, 7 919
XM_017027870.1 XP_016883359.1 6, 7 925
XM_017027871.1 XP_016883360.1 7, 8 1005
GenBank mRNA AF220416.1 - 7, 8 824
AF258787.1 - 6, 7 908
AF348207.1 - 5, 6 847
AK054597.1 - 7, 8 1179
AK314503.1 - 6, 7 886
AY027523.1 - 5, 6 847
AY134747.1 - 6, 7 899
BC006415.1 - 5, 6 782
BC016948.1 - 6, 7 875
BC048210.1 - 7, 8 955
BC080598.1 - 6, 7 886
BC093405.1 - 6, 7 922
BC150178.1 - 6, 7 888 40148074
L09749.1 - 5, 6 801

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001107795, XM_006235404, XM_008762342, FQ232700, FQ228046, and others.

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
histone H4-K5 acetylation
histone H4-K8 acetylation
histone H4-K16 acetylation
regulation of signal transduction by p53 class mediator
DNA binding
protein binding
zinc ion binding
histone acetyltransferase activity (H4-K5 specific)
histone acetyltransferase activity (H4-K8 specific)
histone acetyltransferase activity (H4-K16 specific)

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0