This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other C6ORF203 SIRNA Assays ›
Gene Symbol:
C6orf203
Gene Name:
chromosome 6 open reading frame 203
Gene Aliases:
HSPC230, PRED31
Chromosome Location:
Chr.6: 107028172 - 107051586 on Build GRCh38
UniGene ID:
Hs.486084
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s27761

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C6orf203
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142468.2 NP_001135940.1 2 728
NM_001142470.2 NP_001135942.1 3 811
NM_016487.4 NP_057571.1 2 202
XM_011535876.2 XP_011534178.1 3 306
XM_011535877.2 XP_011534179.1 3 254
XM_017010919.1 XP_016866408.1 2 201
XM_017010920.1 XP_016866409.1 2 694
GenBank mRNA AF151064.1 - 2 158
AK091564.1 - 2 687
BC010899.2 - 1 104 4078296
BX333652.2 - 3 414
DB499205.1 - 2 202

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-hssmq:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline