This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other HEATR5B SIRNA Assays ›
Gene Symbol:
HEATR5B
Gene Name:
HEAT repeat containing 5B
Gene Aliases:
-
Chromosome Location:
Chr.2: 36980999 - 37084384 on Build GRCh38
UniGene ID:
Hs.744084
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s29080

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: HEATR5B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019024.2 NP_061897.1 27 4253
XM_006712034.2 XP_006712097.1 27 4192
XM_006712035.3 XP_006712098.1 27 4256
XM_011532934.2 XP_011531236.1 27 4720
XM_011532935.2 XP_011531237.1 16 2392
XM_017004378.1 XP_016859867.1 27 4192
XM_017004379.1 XP_016859868.1 28 4630
XM_017004380.1 XP_016859869.1 28 4403
GenBank mRNA AB037835.1 - 18 2700
AK026928.1 - 4 513
AK096637.1 - 27 4192
BC085607.1 - 1 25
BC150605.1 - 27 4238 9056975
BX538008.1 - 21 3176
BX648805.1 - 13 2413

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-4spjb:80/100.66.76.244:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0