This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF586 SIRNA Assays ›
Gene Symbol:
ZNF586
Gene Name:
zinc finger protein 586
Gene Aliases:
-
Chromosome Location:
Chr.19: 57769652 - 57780616 on Build GRCh38
UniGene ID:
Hs.625133
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s29479

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF586
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077426.2 NP_001070894.1 2 279
NM_001204814.1 NP_001191743.1 4 456
NM_017652.3 NP_060122.2 3 406
GenBank mRNA AB074270.1 - 3 389
AK000077.1 - 3 388
AK095993.1 - 4 454
AK300419.1 - 2 236
AK302151.1 - 4 362
BC124558.1 - 2 217 40085152
BP352135.1 - 2 280
DA904413.1 - 3 401

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
RNA polymerase II regulatory region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline