This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other IMPAD1 SIRNA Assays ›
Gene Symbol:
IMPAD1
Gene Name:
inositol monophosphatase domain containing 1
Gene Aliases:
GPAPP, IMP 3, IMP-3, IMPA3
Chromosome Location:
Chr.8: 56957929 - 56993874 on Build GRCh38
UniGene ID:
Hs.438689
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s29775

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: IMPAD1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017813.4 NP_060283.3 2, 3 842
GenBank mRNA AK000428.1 - 2, 3 839
AY032885.1 - 2, 3 556
BC017797.1 - 2, 3 791 4306100
BC067814.1 - 2, 3 815 5295820

Back To Top

More Information


Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Function(s) Process(es)

endochondral ossification
chondrocyte development
inositol biosynthetic process
post-embryonic development
dephosphorylation
chondroitin sulfate metabolic process
embryonic digit morphogenesis
phosphatidylinositol phosphorylation
magnesium ion binding
3'-nucleotidase activity
3'(2'),5'-bisphosphate nucleotidase activity
inositol monophosphate 1-phosphatase activity
inositol monophosphate 3-phosphatase activity
inositol monophosphate 4-phosphatase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-s8qkh:80/100.66.76.99:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0