This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CHN2 SIRNA Assays ›
Gene Symbol:
CHN2
Gene Name:
chimerin 2
Gene Aliases:
ARHGAP3, BCH, CHN2-3, RHOGAP3
Chromosome Location:
Chr.7: 29146547 - 29514335 on Build GRCh38
UniGene ID:
Hs.654611
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s3019

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CHN2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039936.2 NP_001035025.1 1 614
NM_001293069.1 NP_001279998.1 8 885
NM_001293070.1 NP_001279999.1 8 1099
NM_001293071.1 NP_001280000.1 6 1004
NM_001293072.1 NP_001280001.1 7 817
NM_001293076.1 NP_001280005.1 1 614
NM_001293077.1 NP_001280006.1 1 614
NM_001293078.1 NP_001280007.1 1 614
NM_001293079.1 NP_001280008.1 1 614
NM_001293080.1 NP_001280009.1 1 614
NM_004067.3 NP_004058.1 7 1060
XM_011515105.2 XP_011513407.1 10 940
XM_011515106.2 XP_011513408.1 9 901
XM_011515107.2 XP_011513409.1 9 1683
XM_017011721.1 XP_016867210.1 11 958
XM_017011722.1 XP_016867211.1 10 1701
GenBank mRNA AK026415.1 - 1 614
AK293959.1 - 8 679
AK294032.1 - 7 817
AK294271.1 - 8 886
AK313021.1 - 7 834
BC112155.1 - 7 805 8327623
EU732754.1 - 1 215
EU732755.1 - 1 215
EU732756.1 - 1 215
EU732757.1 - 1 215
EU732758.1 - 1 215
EU732760.1 - 6 567
EU732762.1 - 4 191
EU732766.1 - 7 623
GQ924106.1 - 8 886
U28926.1 - 7 857

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of signal transduction
intracellular signal transduction
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
SH3/SH2 adaptor activity
GTPase activator activity
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0