siRNA Details
| Gene Symbol: CRTAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206528.2 | NP_001193457.1 | 4 | 807 | ||
| NM_018058.6 | NP_060528.3 | 4 | 807 | |||
| XM_005269938.4 | XP_005269995.1 | 4 | 600 | |||
| XM_011539917.1 | XP_011538219.1 | 4 | 600 | |||
| XM_017016366.1 | XP_016871855.1 | 4 | 600 | |||
| XM_017016367.1 | XP_016871856.1 | 4 | 600 | |||
| GenBank mRNA | AJ276171.1 | - | 4 | 600 | ||
| AJ279016.1 | - | 3 | 427 | |||
| AJ421515.1 | - | 3 | 427 | |||
| AK001182.1 | - | 2 | 139 | |||
| AK057190.1 | - | 4 | 807 | |||
| AK292084.1 | - | 4 | 569 | |||
| BC034245.1 | - | 2 | 69 | 4178363 | ||

CRTAC1
Genomic Map