This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DZANK1 SIRNA Assays ›
Gene Symbol:
DZANK1
Gene Name:
double zinc ribbon and ankyrin repeat domains 1
Gene Aliases:
ANKRD64, C20orf12, C20orf84, bA189K21.1, bA189K21.8, dJ568F9.2
Chromosome Location:
Chr.20: 18383367 - 18467185 on Build GRCh38
UniGene ID:
Hs.472225
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s30380

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 388.00
Your Price
Online offer: 287.12
287.12
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: DZANK1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099407.1 NP_001092877.1 7 804
XM_005260742.3 XP_005260799.1 7 856
XM_005260743.3 XP_005260800.1 6 692
XM_006723584.2 XP_006723647.1 6 731
XM_011529268.2 XP_011527570.1 7 956
XM_011529271.2 XP_011527573.1 7 1569
XM_011529272.2 XP_011527574.1 7 952
XM_011529275.2 XP_011527577.1 6 1442
XM_017027909.1 XP_016883398.1 7 957
XM_017027910.1 XP_016883399.1 7 822
XM_017027911.1 XP_016883400.1 7 818
XM_017027912.1 XP_016883401.1 7 1525
XM_017027913.1 XP_016883402.1 7 1510
XM_017027914.1 XP_016883403.1 7 764
XM_017027915.1 XP_016883404.1 7 760
XM_017027916.1 XP_016883405.1 6 1442
XM_017027917.1 XP_016883406.1 8 1619
XM_017027918.1 XP_016883407.1 8 873
XM_017027919.1 XP_016883408.1 8 869
XM_017027920.1 XP_016883409.1 6 828
GenBank mRNA AK055544.1 - 6 581
AK131451.1 - 8 1443
AK295073.1 - 7 696
AK299746.1 - 7 727
BC036231.1 - 6 777
BC130341.1 - 6 632 40146372
BC144153.1 - 6 632 9052673
CR749327.1 - 8 856

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mitophagy in response to mitochondrial depolarization
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline