This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DZANK1 SIRNA Assays ›
Gene Symbol:
DZANK1
Gene Name:
double zinc ribbon and ankyrin repeat domains 1
Gene Aliases:
ANKRD64, C20orf12, C20orf84, bA189K21.1, bA189K21.8, dJ568F9.2
Chromosome Location:
Chr.20: 18383367 - 18467185 on Build GRCh38
UniGene ID:
Hs.472225
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s30381

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: DZANK1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099407.1 NP_001092877.1 17 1948
XM_005260742.3 XP_005260799.1 17 2000
XM_005260743.3 XP_005260800.1 16 1854
XM_006723584.2 XP_006723647.1 15 1775
XM_011529268.2 XP_011527570.1 17 2118
XM_011529271.2 XP_011527573.1 17 2731
XM_011529272.2 XP_011527574.1 17 2114
XM_011529275.2 XP_011527577.1 16 2604
XM_011529277.2 XP_011527579.1 9 2845
XM_017027909.1 XP_016883398.1 17 2101
XM_017027910.1 XP_016883399.1 17 1984
XM_017027911.1 XP_016883400.1 17 1980
XM_017027912.1 XP_016883401.1 17 2687
XM_017027913.1 XP_016883402.1 17 2672
XM_017027914.1 XP_016883403.1 17 1926
XM_017027915.1 XP_016883404.1 17 1922
XM_017027916.1 XP_016883405.1 15 2486
XM_017027917.1 XP_016883406.1 18 2781
XM_017027918.1 XP_016883407.1 18 2035
XM_017027919.1 XP_016883408.1 18 2031
XM_017027920.1 XP_016883409.1 16 1990
XM_017027921.1 XP_016883410.1 10 1152
GenBank mRNA AK001462.1 - 11 1135
AK026306.1 - 3 157
AK055544.1 - 16 1743
AK131451.1 - 19 2771
AK295073.1 - 17 1858
AK299746.1 - 17 1871
BC130341.1 - 16 1776 40146372
BC144153.1 - 16 1776 9052673
CR749327.1 - 18 2000
DQ104738.1 - 8 869

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mitophagy in response to mitochondrial depolarization
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mlg7h:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline