siRNA Details
| Gene Symbol: PRR34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018280.2 | NP_060750.1 | 2 | 511 | ||
| GenBank mRNA | AK001807.1 | - | 2 | 511 | ||
| AK096201.1 | - | 2 | 163 | |||
| AU134173.1 | - | 2 | 511 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRR34| Gene Symbol: PRR34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018280.2 | NP_060750.1 | 2 | 511 | ||
| GenBank mRNA | AK001807.1 | - | 2 | 511 | ||
| AK096201.1 | - | 2 | 163 | |||
| AU134173.1 | - | 2 | 511 | |||