siRNA Details
| Gene Symbol: ACER3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300953.1 | NP_001287882.1 | 6 | 465 | ||
| NM_001300954.1 | NP_001287883.1 | 7 | 647 | |||
| NM_001300955.1 | NP_001287884.1 | 6 | 523 | |||
| NM_018367.6 | NP_060837.3 | 7 | 576 | |||
| XM_011545151.2 | XP_011543453.1 | 7 | 542 | |||
| XM_011545152.2 | XP_011543454.1 | 7 | 512 | |||
| XM_011545153.2 | XP_011543455.1 | 7 | 542 | |||
| XM_017017988.1 | XP_016873477.1 | 7 | 542 | |||
| GenBank mRNA | AF214454.1 | - | 7 | 530 | ||
| AF327353.1 | - | 7 | 541 | |||
| AK002100.1 | - | 9 | 707 | |||
| AK293800.1 | - | 8 | 547 | |||
| AK294978.1 | - | 6 | 396 | |||
| AK295142.1 | - | 7 | 564 | |||
| AK295327.1 | - | 8 | 660 | |||
| AK308856.1 | - | 9 | 704 | |||
| AK315000.1 | - | 7 | 542 | |||
| AK316120.1 | - | 6 | 461 | |||
| BC063034.1 | - | 8 | 536 | |||
| BC073853.1 | - | 7 | 493 | 3867010 | ||
| BG702017.1 | - | 7 | 581 | |||

ACER3
Genomic Map