This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other HEMGN SIRNA Assays ›
Gene Symbol:
HEMGN
Gene Name:
hemogen
Gene Aliases:
CT155, EDAG, EDAG-1, NDR
Chromosome Location:
Chr.9: 97926791 - 97944915 on Build GRCh38
UniGene ID:
Hs.176626
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s30831

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 392.00
Your Price
Online offer: 380.65
380.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: HEMGN
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018437.4 NP_060907.2 4 1313
NM_197978.2 NP_932095.1 3 1367
XM_005252086.1 XP_005252143.1 4 1275
XM_011518845.2 XP_011517147.1 4 1187
XM_011518846.2 XP_011517148.1 4 1297
GenBank mRNA AF116617.1 - 1 769
AF130060.2 - 3 1100
AF228713.2 - 4 1220
AF306681.1 - 1 191
AF306682.1 - 3 1149
AF306683.1 - 3 1115
AF322875.1 - 4 1219
AK292441.1 - 4 1239
AL833094.1 - 3 1159
AY244805.1 - 3 1313
AY255672.1 - 3 1218
BC048324.1 - 4 1222 5166612

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
cell differentiation
regulation of osteoblast differentiation
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline