This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other C5ORF15 SIRNA Assays ›
Gene Symbol:
C5orf15
Gene Name:
chromosome 5 open reading frame 15
Gene Aliases:
HTGN29, KCT2
Chromosome Location:
Chr.5: 133955507 - 133968715 on Build GRCh38
UniGene ID:
Hs.744034
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s32471

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C5orf15
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020199.2 NP_064584.1 2 408
GenBank mRNA AF226055.1 - 2 481
AK074963.1 - 2 320
AK298515.1 - 2 300
AK315362.1 - 2 306
AY157581.1 - 2 317
BC020875.1 - 2 299 4610607
BP245947.1 - 2 408

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mlg7h:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline