This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TRPC7 SIRNA Assays ›
Gene Symbol:
TRPC7
Gene Name:
transient receptor potential cation channel subfamily C member 7
Gene Aliases:
TRP7
Chromosome Location:
Chr.5: 136213311 - 136365476 on Build GRCh38
UniGene ID:
Hs.591263
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s32703

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TRPC7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167576.1 NP_001161048.1 5 1679
NM_001167577.1 NP_001161049.1 6 1844
NM_020389.2 NP_065122.1 7 2027
GenBank mRNA AJ272034.1 - 6 1805
AJ421783.1 - 6 1805
AJ549088.1 - 5 1622
AJ549089.1 - 5 1640
AJ549090.1 - 3 1240
AY167926.1 - 5 1637
AY167927.1 - 4 1457
BC128185.1 - 6 1805 40113370
BC128186.1 - 6 1821

Back To Top

More Information


Panther Classification:

Molecular Function -

ion channel transporter

Gene Ontology Categories:

Function(s) Process(es)

manganese ion transport
single fertilization
platelet activation
regulation of cytosolic calcium ion concentration
calcium ion transmembrane transport
calcium channel activity
protein binding
store-operated calcium channel activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-m9fgr:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline