siRNA Details
| Gene Symbol: WDR35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006657.1 | NP_001006658.1 | 21 | 2540 | ||
| NM_020779.3 | NP_065830.2 | 20 | 2507 | |||
| XM_011533007.2 | XP_011531309.1 | 10 | 1232 | |||
| GenBank mRNA | AB037757.2 | - | 21 | 2548 | ||
| AK091205.1 | - | 14 | 1875 | |||
| AK122917.1 | - | 20 | 2432 | |||
| AK315094.1 | - | 20 | 2432 | |||
| AL832903.1 | - | 9 | 1037 | |||
| BC036659.2 | - | 20 | 2506 | 5266940 | ||

WDR35
Genomic Map