siRNA Details
| Gene Symbol: KIAA1524 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020890.2 | NP_065941.2 | 16 | 2238 | ||
| XM_006713716.3 | XP_006713779.1 | 16 | 2022 | |||
| XM_006713717.2 | XP_006713780.1 | 17 | 2134 | |||
| XM_011513056.1 | XP_011511358.1 | 16 | 1982 | |||
| XM_011513057.2 | XP_011511359.1 | 17 | 2103 | |||
| GenBank mRNA | AB040957.1 | - | 16 | 1965 | ||
| AF334474.1 | - | 16 | 2037 | |||
| AK022912.1 | - | 7 | 849 | |||
| AK056933.1 | - | 16 | 2047 | |||
| AK308315.1 | - | 16 | 1984 | |||
| AL713689.1 | - | 16 | 2033 | |||
| BC017659.1 | - | 2 | 111 | |||
| BC063433.1 | - | 16 | 2029 | |||
| BC082961.1 | - | 16 | 1991 | |||
| BC130564.1 | - | 16 | 2005 | 40146595 | ||
| BC136371.1 | - | 16 | 2005 | 9020358 | ||

KIAA1524
Genomic Map