This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other USP37 SIRNA Assays ›
Gene Symbol:
USP37
Gene Name:
ubiquitin specific peptidase 37
Gene Aliases:
-
Chromosome Location:
Chr.2: 218450251 - 218568393 on Build GRCh38
UniGene ID:
Hs.166068
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s33613

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: USP37
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020935.2 NP_065986.2 7, 8 1007
XM_005246720.2 XP_005246777.1 7, 8 1079
XM_005246721.3 XP_005246778.1 7, 8 971
XM_005246722.3 XP_005246779.1 7, 8 1011
XM_005246724.1 XP_005246781.1 7, 8 1007
XM_011511538.2 XP_011509840.1 6, 7 952
XM_011511541.2 XP_011509843.1 7, 8 1007
XM_017004577.1 XP_016860066.1 7, 8 965
GenBank mRNA AB046814.1 - 3, 4 452
AK095577.1 - 7, 8 965
AK128583.1 - 7, 8 1011
AL832645.2 - 7, 8 1006
BC050461.1 - 7, 8 976
BC112901.1 - 6, 7 807 6016503
BC133007.1 - 7, 8 892 40147029
BC133009.1 - 7, 8 937 40147031
BC144249.1 - 7, 8 892 9052769
BC144252.1 - 7, 8 932 9052772
BX538024.1 - 7, 8 972

Back To Top

More Information


Panther Classification:

Molecular Function -

cysteine protease

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
ubiquitin-dependent protein catabolic process
mitotic nuclear division
protein deubiquitination
protein K11-linked deubiquitination
cell division
protein K48-linked deubiquitination
cysteine-type endopeptidase activity
thiol-dependent ubiquitin-specific protease activity
protein binding
protein kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0