This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CWC22 SIRNA Assays ›
Gene Symbol:
CWC22
Gene Name:
CWC22 homolog, spliceosome-associated protein
Gene Aliases:
EIF4GL, NCM, fSAPb
Chromosome Location:
Chr.2: 179944876 - 180007359 on Build GRCh38
UniGene ID:
Hs.311363
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s33633

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CWC22
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020943.2 NP_065994.1 10 1427
XM_005246726.2 XP_005246783.1 10 1530
GenBank mRNA AB046824.1 - 9 1246
AK025635.1 - 10 1284
AK026978.1 - 3 384
BC016651.1 - 9 1251
BC031216.1 - 10 1427
BC041662.1 - 3 378
BC053573.1 - 7 949
BC057826.1 - 10 1285
BC093952.1 - 9 1149 7939797
BC093954.1 - 9 1149 7939799
BC143433.1 - 8 1232

Back To Top

More Information


Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
regulation of mRNA splicing, via spliceosome
RNA binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-7wnc4:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline