This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TSKS SIRNA Assays ›
Gene Symbol:
TSKS
Gene Name:
testis specific serine kinase substrate
Gene Aliases:
PPP1R161, STK22S1, TSKS1, TSSKS
Chromosome Location:
Chr.19: 49739753 - 49763328 on Build GRCh38
UniGene ID:
Hs.515858
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s34052

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TSKS
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021733.1 NP_068379.1 5 628
XM_011527201.1 XP_011525503.1 5 698
XM_011527202.1 XP_011525504.1 5 698
XM_011527203.2 XP_011525505.1 5 698
GenBank mRNA BC058862.1 - 5 656 5170446

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of phosphatase activity
protein binding
protein kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-cj8gb:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline