siRNA Details
| Gene Symbol: ANO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001313726.1 | NP_001300655.1 | 14 | 1607 | ||
| NM_001313727.1 | NP_001300656.1 | 10 | 1165 | |||
| NM_031418.3 | NP_113606.2 | 13 | 1515 | |||
| XM_017018118.1 | XP_016873607.1 | 11 | 1143 | |||
| XM_017018119.1 | XP_016873608.1 | 8 | 755 | |||
| GenBank mRNA | AJ300461.1 | - | 13 | 2222 | ||
| AK295143.1 | - | 10 | 1078 | |||
| AK295816.1 | - | 10 | 1077 | |||
| AK302659.1 | - | 13 | 1554 | |||
| AK304781.1 | - | 14 | 1448 | |||
| AK316018.1 | - | 10 | 1077 | |||

ANO3
Genomic Map