siRNA Details
| Gene Symbol: CPN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080513.3 | NP_001073982.3 | 2 | 1048 | ||
| NM_001291988.1 | NP_001278917.1 | 2 | 1045 | |||
| XM_005269280.4 | XP_005269337.1 | 3 | 1177 | |||
| GenBank mRNA | BC031569.1 | - | 2 | 996 | 5188827 | |
| BC042334.1 | - | 2 | 1004 | 5190234 | ||
| BC137398.1 | - | 2 | 1038 | 9021396 | ||
| BC137403.1 | - | 2 | 1038 | 9021401 | ||
| J05158.2 | - | 1 | 962 | |||

CPN2
Genomic Map