This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TMEM108 SIRNA Assays ›
Gene Symbol:
TMEM108
Gene Name:
transmembrane protein 108
Gene Aliases:
CT124
Chromosome Location:
Chr.3: 133038288 - 133397775 on Build GRCh38
UniGene ID:
Hs.191616
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s35339

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 388.00
Your Price
Online offer: 287.12
287.12
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TMEM108
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136469.2 NP_001129941.1 6 1969
NM_001282865.1 NP_001269794.1 4 625
NM_023943.3 NP_076432.1 6 1973
XM_011513097.2 XP_011511399.1 5 3975
XM_011513098.2 XP_011511400.1 7 2068
XM_017007080.1 XP_016862569.1 7 2125
XM_017007081.1 XP_016862570.1 5 500
XM_017007082.1 XP_016862571.1 4 2564
GenBank mRNA AK095050.1 - 4 619
AL136578.1 - 3 1325
AY358381.1 - 6 1855
BC000568.2 - 6 1852 3163990
DB522014.1 - 2 183

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gm74s:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline