This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FASTKD3 SIRNA Assays ›
Gene Symbol:
FASTKD3
Gene Name:
FAST kinase domains 3
Gene Aliases:
-
Chromosome Location:
Chr.5: 7859159 - 7869037 on Build GRCh38
UniGene ID:
Hs.653162
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s35519

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FASTKD3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024091.3 NP_076996.2 2 266
XM_006714498.1 XP_006714561.1 2 177
GenBank mRNA AK026927.1 - 2 231
BC113563.1 - 1 181 8322615
BC143517.1 - 1 181
BC143518.1 - 1 181
BQ921348.1 - 2 223
DA297400.1 - 2 266

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
cellular respiration
protein kinase activity
protein binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-9wwlr:80/100.66.77.227:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline