This product has been added to your favorites list. Go to
My Favorites
Please enter the information below and press OK to send your cart to Core Services for purchase.
Name
Phone number
Email
Budget number
PI Name
PI Email
Building and Room number
Pickup
Delivery
System Message
OK
Cancel
LOADING
See other MFSD11 SIRNA Assays ›
Gene Symbol:
MFSD11
Gene Name:
major facilitator superfamily domain containing 11
Gene Aliases:
ET
Chromosome Location:
Chr.17: 76736565 - 76803805 on Build GRCh38
UniGene ID:
Hs.732019
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s35648
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MFSD11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242532.1
NP_001229461.1
11
1757
NM_001242533.1
NP_001229462.1
12
1641
NM_001242534.1
NP_001229463.1
12
1503
NM_001242535.1
NP_001229464.1
12
1452
NM_001242536.1
NP_001229465.1
10
1485
NM_001242537.1
NP_001229466.1
10
1296
NM_024311.3
NP_077287.1
12
1359
XM_011525236.2
XP_011523538.1
11
1021
XM_011525237.2
XP_011523539.2
12
1464
XM_011525238.2
XP_011523540.2
12
1064
XM_011525239.2
XP_011523541.2
12
1034
XM_011525240.2
XP_011523542.2
12
1060
XM_011525241.2
XP_011523543.2
12
1070
XM_011525242.1
XP_011523544.1
11
1021
XM_011525244.1
XP_011523546.1
11
1021
XM_011525247.2
XP_011523549.2
12
1504
XM_017025065.1
XP_016880554.1
12
1092
XM_017025066.1
XP_016880555.1
12
1034
XM_017025067.1
XP_016880556.1
12
1092
XM_017025068.1
XP_016880557.1
12
1504
XM_017025069.1
XP_016880558.1
10
1192
XM_017025070.1
XP_016880559.1
10
1192
XM_017025071.1
XP_016880560.1
4
381
GenBank mRNA
AF015184.1
-
12
1071
AF015185.1
-
10
1102
AF015186.1
-
12
1215
AK000233.1
-
11
1139
AK025849.1
-
11
894
AK308058.1
-
10
940
BC002753.2
-
12
1159
3629029
BC062563.1
-
12
1252
5140496
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67f85b6989-gm74s:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline