This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other C10ORF76 SIRNA Assays ›
Gene Symbol:
C10orf76
Gene Name:
chromosome 10 open reading frame 76
Gene Aliases:
-
Chromosome Location:
Chr.10: 101845599 - 102056237 on Build GRCh38
UniGene ID:
Hs.16004
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s35876

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C10orf76
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024541.2 NP_078817.2 21, 22 1682
XM_011540151.2 XP_011538453.1 21, 22 1727
XM_011540152.2 XP_011538454.1 20, 21 1614
XM_011540153.2 XP_011538455.1 20, 21 1578
XM_011540154.2 XP_011538456.1 19, 20 1512
XM_011540156.2 XP_011538458.1 14, 15 1646
XM_011540157.2 XP_011538459.1 20, 21 1631
XM_017016635.1 XP_016872124.1 21, 22 1680
XM_017016636.1 XP_016872125.1 21, 22 1680
GenBank mRNA AK023176.1 - 4, 5 311
BC110511.1 - 4, 5 311

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_198296, BC048392, BC058989, BC046389.

Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-j4gfx:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0