This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other VEPH1 SIRNA Assays ›
Gene Symbol:
VEPH1
Gene Name:
ventricular zone expressed PH domain containing 1
Gene Aliases:
MELT, VEPH
Chromosome Location:
Chr.3: 157259742 - 157503626 on Build GRCh38
UniGene ID:
Hs.658046
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s36072

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 388.00
Your Price
Online offer: 374.65
374.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: VEPH1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167911.1 NP_001161383.1 4 726
NM_001167912.1 NP_001161384.1 4 726
NM_001167915.1 NP_001161387.1 5 1073
NM_001167916.1 NP_001161388.1 4 811
NM_001167917.1 NP_001161389.1 3 655
NM_024621.2 NP_078897.2 4 917
XM_011513134.2 XP_011511436.1 3 660
XM_011513135.1 XP_011511437.1 4 556
GenBank mRNA AB051479.1 - 4 726
AK027625.1 - 4 815
AL713656.1 - 3 514
BC057999.1 - 4 814
BC101660.1 - 3 553 8069166
BC111017.1 - 4 718 5174889
BC113555.1 - 3 553 8322607
BG194590.1 - 2 352

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-98cc9b549-wzvtq:80/100.66.78.229:80.
git-commit: 5e7f4b920385fdbbb5f8d978f1c7c2eb3f871195
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.40.1-Offline